Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200016782.mm.1 |
NM_021335.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016782.mm.1 |
NM_021335.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016782.mm.1 |
TC0200002127.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016787.mm.1 |
NM_021335.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016787.mm.1 |
NM_021335.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016787.mm.1 |
TC0200002127.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016784.mm.1 |
NM_021335.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016784.mm.1 |
NM_021335.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016784.mm.1 |
TC0200002127.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016789.mm.1 |
NM_021335.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016789.mm.1 |
NM_021335.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016789.mm.1 |
TC0200002127.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016777.mm.1 |
NM_021335.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016777.mm.1 |
NM_021335.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016777.mm.1 |
TC0200002127.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016788.mm.1 |
NM_021335.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016788.mm.1 |
NM_021335.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016788.mm.1 |
TC0200002127.mm.1 |
MTA TC |