Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085449.1 | NP_001078918.1 | 15 - 16 | 2814 | 97 | |
NM_001085450.1 | NP_001078919.1 | 17 - 18 | 3121 | 97 | ||
NM_007615.4 | NP_031641.2 | 18 - 19 | 3239 | 97 | ||
GenBank mRNA | AK168453.1 | - | 15 - 16 | 2812 | 97 | |
BC043108.1 | - | 17 - 18 | 3056 | 6406902 | 97 | |
BC046589.1 | - | 14 - 15 | 3157 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |