Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172666.3 | NP_766254.2 | 6 - 7 | 782 | 113 | |
GenBank mRNA | AK031049.1 | - | 6 - 7 | 784 | 113 | |
AK031154.1 | - | 6 - 7 | 623 | 113 | ||
AK042745.1 | - | 6 - 7 | 622 | 113 | ||
AK053686.1 | - | 6 - 7 | 771 | 113 | ||
AK079447.1 | - | 5 - 6 | 462 | 113 | ||
BC063086.1 | - | 6 - 7 | 767 | 6332574 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |