Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172666.3 | NP_766254.2 | 7 - 8 | 866 | 92 | |
GenBank mRNA | AK031049.1 | - | 7 - 8 | 868 | 92 | |
AK031154.1 | - | 7 - 8 | 707 | 92 | ||
AK042745.1 | - | 7 - 8 | 706 | 92 | ||
AK053686.1 | - | 7 - 8 | 855 | 92 | ||
AK079447.1 | - | 6 - 7 | 546 | 92 | ||
BC063086.1 | - | 7 - 8 | 851 | 6332574 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |