Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144824.2 | NP_659073.1 | 1 - 2 | 656 | 58 | |
GenBank mRNA | AK077570.1 | - | 1 - 2 | 559 | 58 | |
AK145219.1 | - | 1 - 2 | 582 | 58 | ||
AK169271.1 | - | 1 - 2 | 559 | 58 | ||
BC021790.1 | - | 1 - 2 | 656 | 4220923 | 58 | |
BC069868.1 | - | 2 - 3 | 700 | 5101220 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |