Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK036520.1 | - | 13 - 14 | 1322 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |