Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029687.3 | NP_083963.1 | 7 - 8 | 867 | 85 | |
GenBank mRNA | AK007246.1 | - | 7 - 8 | 858 | 85 | |
AK077027.1 | - | 1115 | 85 | |||
BC115629.1 | - | 7 - 8 | 845 | 40102866 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |