Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114087.2 | NP_001107559.1 | 6 - 7 | 626 | 97 | |
NM_026131.4 | NP_080407.3 | 6 - 7 | 524 | 97 | ||
NR_104287.1 | 7 - 8 | 643 | 97 | |||
GenBank mRNA | AK003019.1 | - | 7 - 8 | 600 | 97 | |
AK003338.1 | - | 6 - 7 | 515 | 97 | ||
AK010141.1 | - | 6 - 7 | 513 | 97 | ||
AK077998.1 | - | 6 - 7 | 618 | 97 | ||
BC049565.1 | - | 6 - 7 | 512 | 6588160 | 97 | |
BC052698.1 | - | 6 - 7 | 597 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |