Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033610.2 | NP_291088.1 | 3 - 4 | 305 | 79 | |
GenBank mRNA | BC019409.1 | - | 3 - 4 | 305 | 4482283 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |