Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290782.1 | NP_001277711.1 | 4 - 5 | 605 | 88 | |
NM_183220.3 | NP_899043.1 | 4 - 5 | 660 | 88 | ||
GenBank mRNA | AK011867.1 | - | 4 - 5 | 509 | 88 | |
AK040509.1 | - | 4 - 5 | 502 | 88 | ||
BC039569.1 | - | 4 - 5 | 637 | 1449080 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |