Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290782.1 | NP_001277711.1 | 5 - 6 | 670 | 55 | |
NM_183220.3 | NP_899043.1 | 5 - 6 | 725 | 55 | ||
GenBank mRNA | BC039569.1 | - | 5 - 6 | 702 | 1449080 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |