Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033452.4 | NP_001028624.2 | 6 - 7 | 953 | 78 | |
GenBank mRNA | AK135828.1 | - | 6 - 7 | 900 | 78 | |
AK142866.1 | - | 4 - 5 | 444 | 78 | ||
AK163963.1 | - | 6 - 7 | 1035 | 78 | ||
BC141227.1 | - | 4 - 5 | 332 | 9055839 | 78 | |
BC145551.1 | - | 4 - 5 | 332 | 78 | ||
BC145552.1 | - | 4 - 5 | 332 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |