Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luzp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178705.5 | NP_848820.4 | 5 - 6 | 1107 | 80 | |
GenBank mRNA | AK032204.1 | - | 5 - 6 | 764 | 80 | |
AK034077.1 | - | 5 - 6 | 725 | 80 | ||
AK034240.1 | - | 5 - 6 | 759 | 80 | ||
AK082126.1 | - | 5 - 6 | 855 | 80 | ||
BC075646.1 | - | 5 - 6 | 737 | 30620475 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |