Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177383.4 | NP_796357.1 | 1 - 2 | 74 | 72 | |
GenBank mRNA | AK048682.1 | - | 1 - 2 | 75 | 72 | |
AK139361.1 | - | 1 - 2 | 75 | 72 | ||
AK141999.1 | - | 108 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Mus musculus Probe spans exons |