Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043335.1 | NP_001036800.1 | 6 - 7 | 1050 | 81 | |
NM_001043336.2 | NP_001036801.1 | 6 - 7 | 667 | 81 | ||
NM_001286346.1 | NP_001273275.1 | 6 - 7 | 804 | 81 | ||
NM_001286347.1 | NP_001273276.1 | 7 - 8 | 718 | 81 | ||
GenBank mRNA | AK044662.1 | - | 6 - 7 | 789 | 81 | |
BC053094.1 | - | 6 - 7 | 884 | 6400458 | 81 | |
BC059839.1 | - | 6 - 7 | 645 | 6417403 | 81 | |
BC079582.1 | - | 6 - 7 | 703 | 5702743 | 81 | |
BC098329.1 | - | 5 - 6 | 446 | 81 | ||
BC125289.1 | - | 7 - 8 | 689 | 40129804 | 81 | |
BC144757.1 | - | 7 - 8 | 689 | 81 | ||
BC150975.1 | - | 6 - 7 | 804 | 9087888 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |