Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200040660.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040660.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040660.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040665.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040665.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040665.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040666.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040666.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040666.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040675.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040675.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040675.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040653.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040653.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040653.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040656.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040656.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040656.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040657.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040657.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040657.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040673.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040673.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040673.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040668.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040668.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040668.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040662.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040662.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040662.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040670.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040670.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040670.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040674.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040674.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040674.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040669.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040669.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040669.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040671.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040671.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040671.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040672.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040672.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040672.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040654.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040654.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040654.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040658.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040658.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040658.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040659.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040659.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040659.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040652.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040652.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040652.mm.1 |
TC0200004750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040676.mm.1 |
NM_024281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040676.mm.1 |
NM_024281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040676.mm.1 |
TC0200004750.mm.1 |
MTA TC |