Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK036623.1 | - | 15 - 16 | 2023 | 67 | |
AK151178.1 | - | 15 - 16 | 1957 | 67 | ||
AK153039.1 | - | 15 - 16 | 1957 | 67 | ||
AK169948.1 | - | 15 - 16 | 2024 | 67 | ||
AY753194.1 | - | 13 - 14 | 1713 | 67 | ||
BC028998.1 | - | 15 - 16 | 1953 | 5356085 | 67 | |
DQ295832.1 | - | 13 - 14 | 1725 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |