Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ippk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276399.1 | NP_001263328.1 | 6 - 7 | 743 | 108 | |
NM_199056.3 | NP_951011.1 | 6 - 7 | 662 | 108 | ||
NR_075085.1 | 7 - 8 | 747 | 108 | |||
GenBank mRNA | AK029085.1 | - | 6 - 7 | 609 | 108 | |
AK031987.1 | - | 6 - 7 | 744 | 107 | ||
AK160092.1 | - | 6 - 7 | 609 | 108 | ||
AK167467.1 | - | 6 - 7 | 637 | 108 | ||
BC024630.1 | - | 7 - 8 | 692 | 4222376 | 108 | |
BC062167.1 | - | 6 - 7 | 574 | 6517021 | 108 | |
BC065157.1 | - | 6 - 7 | 577 | 6416865 | 108 | |
CD353694.1 | - | 3 - 4 | 288 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |