Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007603.3 | NP_031629.3 | 1 - 2 | 205 | 156 | |
GenBank mRNA | AK048028.1 | - | 1 - 2 | 126 | 156 | |
AK145116.1 | - | 1 - 2 | 220 | 168 | ||
CN700442.1 | - | 1 - 2 | 205 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mus musculus Probe spans exons |