Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008569.2 | NP_032595.2 | 11 - 12 | 1621 | 102 | |
GenBank mRNA | AK052404.1 | - | 11 - 12 | 1602 | 102 | |
AK054305.1 | - | 11 - 12 | 1603 | 102 | ||
AK077847.1 | - | 11 - 12 | 1622 | 102 | ||
AK090134.1 | - | 11 - 12 | 1604 | 102 | ||
AK149762.1 | - | 11 - 12 | 1593 | 102 | ||
AK164804.1 | - | 11 - 12 | 1607 | 102 | ||
BC139002.1 | - | 11 - 12 | 1602 | 8862024 | 102 | |
BC145492.1 | - | 11 - 12 | 1602 | 9054055 | 102 | |
X80169.1 | - | 11 - 12 | 1606 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |