Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008569.2 | NP_032595.2 | 4 - 5 | 693 | 129 | |
GenBank mRNA | AK052404.1 | - | 4 - 5 | 674 | 129 | |
AK054305.1 | - | 4 - 5 | 675 | 129 | ||
AK077847.1 | - | 4 - 5 | 694 | 129 | ||
AK090134.1 | - | 4 - 5 | 676 | 129 | ||
AK149762.1 | - | 4 - 5 | 665 | 129 | ||
AK164804.1 | - | 4 - 5 | 679 | 129 | ||
BC139002.1 | - | 4 - 5 | 674 | 8862024 | 129 | |
BC145492.1 | - | 4 - 5 | 674 | 9054055 | 129 | |
X80169.1 | - | 4 - 5 | 678 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |