Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mertk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008587.1 | NP_032613.1 | 4 - 5 | 793 | 62 | |
GenBank mRNA | U21301.1 | - | 4 - 5 | 793 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mus musculus Probe spans exons |