Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181589.3 | NP_853620.1 | 1 - 2 | 110 | 103 | |
GenBank mRNA | AK153766.1 | - | 1 - 2 | 69 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |