Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001110228.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001110228.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001110229.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001110229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001110230.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001110230.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001110231.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001110231.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001110232.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001110232.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001160292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001160292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001160293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_001160293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_010160.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
NM_010160.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024083.mm.1 |
TC0200002894.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024114.mm.1 |
NM_001110231.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024114.mm.1 |
NM_001110231.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024114.mm.1 |
NM_001110232.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024114.mm.1 |
NM_001110232.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024114.mm.1 |
TC0200002894.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024109.mm.1 |
NM_001110229.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024109.mm.1 |
NM_001110229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024109.mm.1 |
NM_001110230.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024109.mm.1 |
NM_001110230.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024109.mm.1 |
NM_001160292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024109.mm.1 |
NM_001160292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024109.mm.1 |
TC0200002894.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024110.mm.1 |
NM_001110229.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024110.mm.1 |
NM_001110229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024110.mm.1 |
NM_001110230.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024110.mm.1 |
NM_001110230.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024110.mm.1 |
NM_001160292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024110.mm.1 |
NM_001160292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024110.mm.1 |
TC0200002894.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024111.mm.1 |
NM_001110229.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024111.mm.1 |
NM_001110229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024111.mm.1 |
NM_001110230.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024111.mm.1 |
NM_001110230.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024111.mm.1 |
NM_001160292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024111.mm.1 |
NM_001160292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024111.mm.1 |
TC0200002894.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024118.mm.1 |
NM_001110232.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024118.mm.1 |
NM_001110232.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024118.mm.1 |
TC0200002894.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001110228.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001110228.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001110229.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001110229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001110230.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001110230.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001110231.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001110231.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001110232.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001110232.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001160292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001160292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001160293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_001160293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_010160.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
NM_010160.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024084.mm.1 |
TC0200002894.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024119.mm.1 |
NM_001110231.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024119.mm.1 |
NM_001110231.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024119.mm.1 |
TC0200002894.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024104.mm.1 |
NM_001110229.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024104.mm.1 |
NM_001110229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024104.mm.1 |
NM_001110230.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024104.mm.1 |
NM_001110230.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024104.mm.1 |
NM_001110231.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024104.mm.1 |
NM_001110231.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024104.mm.1 |
NM_001110232.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024104.mm.1 |
NM_001110232.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024104.mm.1 |
NM_001160292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024104.mm.1 |
NM_001160292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024104.mm.1 |
TC0200002894.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024116.mm.1 |
NM_001110231.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024116.mm.1 |
NM_001110231.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024116.mm.1 |
NM_001110232.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024116.mm.1 |
NM_001110232.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024116.mm.1 |
TC0200002894.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024117.mm.1 |
NM_001110231.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024117.mm.1 |
NM_001110231.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024117.mm.1 |
NM_001110232.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024117.mm.1 |
NM_001110232.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024117.mm.1 |
TC0200002894.mm.1 |
MTA TC |