Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021398.3 | NP_067373.2 | 10 - 11 | 1474 | 83 | |
GenBank mRNA | AF188622.1 | - | 10 - 11 | 1468 | 83 | |
BU516072.1 | - | 4 - 5 | 510 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mus musculus Probe spans exons |