Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081132.1 | NP_001074601.1 | 6 - 7 | 1750 | 83 | |
GenBank mRNA | BC138358.1 | - | 6 - 7 | 1713 | 8861378 | 83 |
BC138359.1 | - | 6 - 7 | 1713 | 8861379 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |