Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146251.4 | NP_666363.3 | 3 - 4 | 449 | 96 | |
GenBank mRNA | AK038505.1 | - | 3 - 4 | 335 | 96 | |
AK088362.1 | - | 3 - 4 | 395 | 96 | ||
AK148380.1 | - | 3 - 4 | 423 | 96 | ||
AK170417.1 | - | 3 - 4 | 395 | 96 | ||
BC021312.1 | - | 3 - 4 | 305 | 96 | ||
BC141365.1 | - | 3 - 4 | 233 | 9055978 | 96 | |
BC141366.1 | - | 3 - 4 | 233 | 9055979 | 96 | |
EU293208.1 | - | 2 - 3 | 120 | 96 | ||
EU368675.1 | - | 2 - 3 | 120 | 96 | ||
EU402948.2 | - | 2 - 3 | 120 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |