Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029983.2 | NP_001025154.1 | 6 - 7 | 908 | 122 | |
GenBank mRNA | AK088523.1 | - | 1 - 2 | 194 | 122 | |
AK214534.1 | - | 1 - 2 | 77 | 122 | ||
AK217415.1 | - | 1 - 2 | 103 | 122 | ||
BC006645.1 | - | 4 - 5 | 524 | 122 | ||
BC138550.1 | - | 6 - 7 | 864 | 8861570 | 122 | |
BC138551.1 | - | 6 - 7 | 864 | 8861571 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |