Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081260.2 | NP_001074729.1 | 7 - 8 | 4772 | 68 | |
GenBank mRNA | AK129436.1 | - | 3 - 4 | 3465 | 68 | |
BC024499.1 | - | 2 - 3 | 670 | 68 | ||
BC025943.1 | - | 2 - 3 | 2221 | 68 | ||
BC058639.1 | - | 4 - 5 | 3916 | 68 | ||
BC150947.1 | - | 7 - 8 | 4692 | 9087858 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |