Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016716.4 | NP_057925.1 | 9 - 10 | 1497 | 128 | |
GenBank mRNA | AF129738.1 | - | 9 - 10 | 1387 | 128 | |
AK129174.1 | - | 10 - 11 | 1885 | 128 | ||
BC027304.1 | - | 9 - 10 | 1522 | 3981170 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |