Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200002232.mm.1 |
NM_175286.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002232.mm.1 |
NM_175286.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002232.mm.1 |
TC0200000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002241.mm.1 |
NM_175286.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002241.mm.1 |
NM_175286.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002241.mm.1 |
TC0200000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002238.mm.1 |
NM_175286.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002238.mm.1 |
NM_175286.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002238.mm.1 |
TC0200000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002240.mm.1 |
NM_175286.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002240.mm.1 |
NM_175286.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002240.mm.1 |
TC0200000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002236.mm.1 |
NM_175286.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002236.mm.1 |
NM_175286.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002236.mm.1 |
TC0200000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002233.mm.1 |
NM_175286.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002233.mm.1 |
NM_175286.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002233.mm.1 |
TC0200000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002234.mm.1 |
NM_175286.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002234.mm.1 |
NM_175286.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002234.mm.1 |
TC0200000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002237.mm.1 |
NM_175286.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002237.mm.1 |
NM_175286.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002237.mm.1 |
TC0200000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002231.mm.1 |
NM_175286.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002231.mm.1 |
NM_175286.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002231.mm.1 |
TC0200000405.mm.1 |
MTA TC |