Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kynu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289593.1 | NP_001276522.1 | 7 - 8 | 676 | 71 | |
NM_001289594.1 | NP_001276523.1 | 7 - 8 | 676 | 71 | ||
NM_027552.2 | NP_081828.1 | 7 - 8 | 676 | 71 | ||
GenBank mRNA | AK014484.1 | - | 7 - 8 | 672 | 71 | |
AK040584.1 | - | 7 - 8 | 651 | 71 | ||
AK050024.1 | - | 7 - 8 | 673 | 71 | ||
BC026950.1 | - | 7 - 8 | 647 | 5136970 | 71 | |
BC069848.1 | - | 7 - 8 | 682 | 5097145 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |