Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016719.1 | NP_057928.1 | 2 - 3 | 435 | 77 | |
GenBank mRNA | AF155647.1 | - | 2 - 3 | 435 | 77 | |
AK010849.1 | - | 2 - 3 | 452 | 77 | ||
AK010903.1 | - | 2 - 3 | 452 | 77 | ||
AK147041.1 | - | 2 - 3 | 444 | 77 | ||
BC138581.1 | - | 2 - 3 | 341 | 8861601 | 77 | |
BC145913.1 | - | 2 - 3 | 341 | 40131216 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |