Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013830.2 | NP_038858.2 | 11 - 12 | 2623 | 119 | |
GenBank mRNA | AB516304.1 | - | 11 - 12 | 2475 | 119 | |
AF033663.1 | - | 10 - 11 | 1913 | 119 | ||
AF283466.1 | - | 11 - 12 | 2621 | 119 | ||
AK076020.1 | - | 3 - 4 | 433 | 119 | ||
AK083926.1 | - | 11 - 12 | 2507 | 119 | ||
BC003769.1 | - | 8 - 9 | 1000 | 119 | ||
BC042521.1 | - | 12 - 13 | 1905 | 119 | ||
BC080864.1 | - | 11 - 12 | 2594 | 119 | ||
BC141272.1 | - | 11 - 12 | 2549 | 9055884 | 119 | |
BC141273.1 | - | 11 - 12 | 2549 | 9055885 | 119 | |
U48737.1 | - | 9 - 10 | 1180 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |