Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028716.4 | NP_082992.1 | 12 - 13 | 1260 | 110 | |
GenBank mRNA | AK014380.1 | - | 12 - 13 | 1261 | 110 | |
BC027776.1 | - | 12 - 13 | 1255 | 5352251 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |