Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009421.3 | NP_033447.2 | 5 - 6 | 601 | 74 | |
GenBank mRNA | AK029135.1 | - | 5 - 6 | 603 | 74 | |
AK089281.1 | - | 4 - 5 | 345 | 74 | ||
AK142931.1 | - | 4 - 5 | 360 | 74 | ||
AK145075.1 | - | 176 | 74 | |||
AK169515.1 | - | 5 - 6 | 394 | 74 | ||
AK172598.1 | - | 1 - 2 | 205 | 74 | ||
BC096595.1 | - | 1 - 2 | 167 | 30748947 | 74 | |
L35302.1 | - | 4 - 5 | 347 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |