Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfmbt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198808.1 | NP_001185737.1 | 9 - 10 | 1162 | 94 | |
NM_177386.5 | NP_796360.2 | 10 - 11 | 1285 | 94 | ||
GenBank mRNA | AK052339.1 | - | 9 - 10 | 1228 | 94 | |
AK147557.1 | - | 9 - 10 | 1167 | 94 | ||
AK147651.1 | - | 10 - 11 | 1140 | 94 | ||
AK220408.1 | - | 7 - 8 | 1027 | 94 | ||
BC139354.1 | - | 9 - 10 | 1073 | 8862376 | 94 | |
BC145610.1 | - | 8 - 9 | 1029 | 9054176 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |