Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200011800.mm.1 |
NM_144783.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011800.mm.1 |
NM_144783.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011800.mm.1 |
TC0200001576.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200011808.mm.1 |
NM_144783.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011808.mm.1 |
NM_144783.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011808.mm.1 |
TC0200001576.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200011803.mm.1 |
NM_144783.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011803.mm.1 |
NM_144783.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011803.mm.1 |
TC0200001576.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200011802.mm.1 |
NM_144783.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011802.mm.1 |
NM_144783.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011802.mm.1 |
TC0200001576.mm.1 |
MTA TC |