Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153587.2 | NP_705815.1 | 3 - 4 | 528 | 104 | |
GenBank mRNA | AK046319.1 | - | 3 - 4 | 305 | 104 | |
AK135786.1 | - | 436 | 104 | |||
AY341873.1 | - | 3 - 4 | 517 | 104 | ||
AY341881.1 | - | 3 - 4 | 517 | 104 | ||
BC035298.1 | - | 3 - 4 | 528 | 4021865 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |