Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289572.1 | NP_001276501.1 | 4 - 5 | 454 | 133 | |
NM_001289574.1 | NP_001276503.1 | 5 - 6 | 616 | 133 | ||
NM_178914.4 | NP_849245.2 | 5 - 6 | 550 | 133 | ||
GenBank mRNA | AK036359.1 | - | 5 - 6 | 534 | 133 | |
AK077500.1 | - | 5 - 6 | 461 | 133 | ||
AK161308.1 | - | 4 - 5 | 400 | 133 | ||
BC046960.1 | - | 5 - 6 | 508 | 6391450 | 133 | |
DQ196101.1 | - | 4 - 5 | 280 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |