Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146199.1 | NP_001139671.1 | 13 - 14 | 2735 | 89 | |
NM_011877.2 | NP_036007.2 | 13 - 14 | 2870 | 89 | ||
GenBank mRNA | BC049893.1 | - | 11 - 12 | 2617 | 89 | |
BC055942.1 | - | 1 - 2 | 1259 | 89 | ||
BC098500.1 | - | 13 - 14 | 2735 | 30081068 | 89 | |
D37801.1 | - | 13 - 14 | 2819 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |