Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172737.4 | NP_766325.3 | 4 - 5 | 1135 | 106 | |
GenBank mRNA | AK085127.1 | - | 3 - 4 | 357 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |