Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010973.2 | NP_001010973.1 | 2 - 3 | 1574 | 84 | |
GenBank mRNA | AK020259.1 | - | 2 - 3 | 1575 | 84 | |
AK163760.1 | - | 2 - 3 | 1571 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mus musculus Probe spans exons |