Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011917.2 | NP_036047.2 | 12 - 13 | 1184 | 120 | |
GenBank mRNA | AK004800.1 | - | 12 - 13 | 1159 | 120 | |
AK011711.1 | - | 12 - 13 | 1182 | 120 | ||
AK031247.1 | - | 12 - 13 | 1227 | 120 | ||
AK053643.1 | - | 12 - 13 | 1177 | 120 | ||
AK133654.1 | - | 12 - 13 | 1166 | 120 | ||
AK163899.1 | - | 11 - 12 | 1237 | 120 | ||
AK167019.1 | - | 12 - 13 | 1204 | 120 | ||
AK168037.1 | - | 12 - 13 | 1230 | 120 | ||
BC004028.1 | - | 4 - 5 | 355 | 3495063 | 120 | |
BC052823.1 | - | 12 - 13 | 1045 | 120 | ||
BC054743.1 | - | 12 - 13 | 1184 | 6400974 | 120 | |
D38517.1 | - | 12 - 13 | 1122 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |