Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK077475.1 | - | 8 - 9 | 881 | 101 | |
AK154760.1 | - | 8 - 9 | 1042 | 101 | ||
AK161339.1 | - | 3 - 4 | 332 | 101 | ||
BC065140.1 | - | 8 - 9 | 915 | 101 | ||
BC086671.1 | - | 8 - 9 | 919 | 30635559 | 101 | |
D38557.1 | - | 8 - 9 | 989 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |