Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080462.2 | NP_536710.1 | 4 - 5 | 375 | 132 | |
GenBank mRNA | AB070524.1 | - | 4 - 5 | 371 | 132 | |
AB444950.1 | - | 3 - 4 | 305 | 132 | ||
AB444951.1 | - | 3 - 4 | 305 | 132 | ||
AF297037.1 | - | 4 - 5 | 375 | 132 | ||
AK050129.1 | - | 4 - 5 | 398 | 132 | ||
AK160131.1 | - | 3 - 4 | 319 | 132 | ||
BC033928.1 | - | 4 - 5 | 367 | 1546890 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |