Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010582.3 | NP_034712.2 | 15 - 16 | 2075 | 81 | |
GenBank mRNA | AK133782.1 | - | 15 - 16 | 1974 | 81 | |
BC034341.1 | - | 15 - 16 | 2049 | 4211576 | 81 | |
X70392.1 | - | 15 - 16 | 2061 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |