Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010582.3 | NP_034712.2 | 8 - 9 | 985 | 92 | |
GenBank mRNA | AK133782.1 | - | 8 - 9 | 884 | 92 | |
AK149533.1 | - | 9 - 10 | 1030 | 92 | ||
BC034341.1 | - | 8 - 9 | 959 | 4211576 | 92 | |
X70392.1 | - | 8 - 9 | 971 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |