Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009369.4 | NP_033395.1 | 2 - 3 | 271 | 73 | |
GenBank mRNA | AK084431.1 | - | 2 - 3 | 289 | 73 | |
AK135536.1 | - | 2 - 3 | 316 | 73 | ||
AK142323.1 | - | 2 - 3 | 291 | 73 | ||
AK150449.1 | - | 2 - 3 | 289 | 73 | ||
AK150833.1 | - | 2 - 3 | 289 | 73 | ||
AK150969.1 | - | 2 - 3 | 290 | 73 | ||
AK151680.1 | - | 2 - 3 | 286 | 73 | ||
AK152365.1 | - | 2 - 3 | 290 | 73 | ||
AK152662.1 | - | 2 - 3 | 289 | 73 | ||
AK155572.1 | - | 2 - 3 | 286 | 73 | ||
AK155828.1 | - | 2 - 3 | 286 | 73 | ||
AK159711.1 | - | 2 - 3 | 286 | 73 | ||
AK164373.1 | - | 2 - 3 | 289 | 73 | ||
AK169912.1 | - | 2 - 3 | 291 | 73 | ||
AK170495.1 | - | 2 - 3 | 286 | 73 | ||
AK171557.1 | - | 1 - 2 | 110 | 73 | ||
BC129900.1 | - | 2 - 3 | 264 | 40111549 | 73 | |
BC129901.1 | - | 2 - 3 | 264 | 40111550 | 73 | |
L19932.1 | - | 2 - 3 | 272 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |