Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011648.5 | NP_035778.3 | 9 - 10 | 979 | 72 | |
GenBank mRNA | AK014519.1 | - | 9 - 10 | 978 | 72 | |
AK029084.1 | - | 9 - 10 | 945 | 72 | ||
BC086691.1 | - | 9 - 10 | 908 | 30682686 | 72 | |
BC092523.1 | - | 9 - 10 | 921 | 30650002 | 72 | |
U02601.1 | - | 9 - 10 | 961 | 72 | ||
U02602.1 | - | 9 - 10 | 961 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |